missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEC Recombinant Human Protein
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain.
Brand: Invitrogen™ PR4801B
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TEC is highly expressed in many hematopoietic cell lines.
Specifications
NP_003206 | |
20mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
TEC, PSCTK4 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
76.4kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
TEC Kinase Family | |
Recombinant | |
Baculovirus |