Learn More
TdT Monoclonal Antibody (19-3), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-5846-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: This 19-3 monoclonal antibody recognizes human and mouse terminal-deoxynucleotidyl transferase (TdT), a 60 kDa polymerase responsible for the template-independent addition of N-nucleotides at gene segment junctions of developing lymphocytes. Expression is found in the nucleus of immature lymphocytes but not in mature lymphocytes or non-lymphoid cells. Tdt plays a critical role during TCR and Ig gene rearrangement. Regulation of expression in the mouse thymus correlates with T cell selection; decreased or absent after positive selection. Additionally Tdt has been shown to be present in lymphoma or lymphoblastic leukemia cells. Applications Reported: This 19-3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 19-3 antibody has been tested by intracellular staining using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5521) followed by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.
Terminal Deoxynucleotidyl Transferase (TdT) is a DNA polymerase located in the cell nucleus which catalyses the polymerization of deoxynucleotides at the 3' hydroxyl ends of oligo or polydeoxynucleotide initiators and functions without a template. TdT is considered to be a highly specific marker for the diagnosis and classification of acute lymphoblastic lymphoma/leuksemias. The determination of TdT expression is most valuable when it is different to differentiate histologically between lymphoblastic lymphoma and Burkitt's lymphoma.Specifications
TdT | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P04053, P09838 | |
DNTT | |
Affinity chromatography | |
RUO | |
1791, 21673 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
19-3 | |
PE | |
DNTT | |
Deoxynucleotidyltransferase terminal; deoxynucleotidyltransferase, terminal; DNA nucleotidylexotransferase; Dntt; Nucleosidetriphosphate DNA Deoxynucleotidylexotransferase; nucleosidetriphosphate:DNA deoxynucleotidylexotransferase; TDT; Terminal addition enzyme; terminal deoxynucleotidyl transferase; terminal deoxynucleotidyltransferase; terminal deoxyribonucleotidyltransferase; terminal transferase | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b κ |
For Research Use Only.