Learn More
Tocris Bioscience™ TCS PIM-1 4a
Selective, ATP-competitive Pim kinase inhibitor
Brand:  Tocris Bioscience™ 371450
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13433474
  • £229.32 / 50mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
Selective, ATP-competitive Pim kinase inhibitor (IC50 values are 24 and 100 nM for Pim-1 and Pim-2 respectively). Displays selectivity over a panel of ∽50 other kinases tested. Exhibits cytotoxicity in PC3 prostate carcinoma cells in vitro (IC50 = 17 μM).
Specifications
Specifications
TCS PIM-1 4a
>98
50mg
TCS PIM-1 4a
273.23