missing translation for 'onlineSavingsMsg'
Learn More

TCS PIM-1 1, Tocris Bioscience™

Selective, ATP-competitive Pim-1 kinase inhibitor

Brand:  Tocris (Bio-Techne) 297950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13442184

  • £445.92 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATP-competitive Pim-1 kinase inhibitor that displays selectivity over Pim-2 and MEK1/2 (IC50 values are 50, > 20000 and > 20000 nM for Pim-1, Pim-2 and MEK1/2 respectively).
Specifications

Chemical Identifiers

491871-58-0
367.202
tcs pim-1 1, 6-5-bromo-2-hydroxyphenyl-2-oxo-4-phenyl-1,2-dihydropyridine-3-carbonitrile, 3-cyano-4-phenyl-6-3-bromo-6-hydroxy phenyl-2 1h-pyridone, 6-5-bromo-2-hydroxyphenyl-2-oxo-4-phenyl-1h-pyridine-3-carbonitrile, 2obj, pim1i, pyridone-based compound, 1, tcs pim-11, pim1-in-1 tcs pim-1 1, 3-cyano-4-phenyl-6-3-bromo-6-hydroxyphenyl-2-pyridone
CHEBI:86453
C1=CC=C(C=C1)C2=C(C(=O)NC(=C2)C3=C(C=CC(=C3)Br)O)C#N
C18H11BrN2O2
SVSYJTYGPLVUOZ-UHFFFAOYSA-N
1235170
6-(5-bromo-2-hydroxyphenyl)-2-oxo-4-phenyl-1H-pyridine-3-carbonitrile

Specifications

TCS PIM-1 1
491871-58-0
tcs pim-1 1, 6-5-bromo-2-hydroxyphenyl-2-oxo-4-phenyl-1,2-dihydropyridine-3-carbonitrile, 3-cyano-4-phenyl-6-3-bromo-6-hydroxy phenyl-2 1h-pyridone, 6-5-bromo-2-hydroxyphenyl-2-oxo-4-phenyl-1h-pyridine-3-carbonitrile, 2obj, pim1i, pyridone-based compound, 1, tcs pim-11, pim1-in-1 tcs pim-1 1, 3-cyano-4-phenyl-6-3-bromo-6-hydroxyphenyl-2-pyridone
C1=CC=C(C=C1)C2=C(C(=O)NC(=C2)C3=C(C=CC(=C3)Br)O)C#N
367.202
CHEBI:86453
>98%
50mg
C18H11BrN2O2
SVSYJTYGPLVUOZ-UHFFFAOYSA-N
6-(5-bromo-2-hydroxyphenyl)-2-oxo-4-phenyl-1H-pyridine-3-carbonitrile
1235170
367.2
Videos