missing translation for 'onlineSavingsMsg'
Learn More

TCS JNK 6o, Tocris Bioscience™

Selective JNK inhibitor

Brand:  Tocris (Bio-Techne) 322210

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13462594

  • £188.76 / 10mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

ATP-competitive c-Jun N-terminal kinase (JNK) inhibitor (IC50 values are 2, 4 and 52 nM for JNK1, JNK2 and JNK3 respectively). Displays > 1000 fold selectivity over other kinases, including ERK2 and p38. Inhibits c-Jun phosphorylation (EC50 = 920 nM) and prevents collagen-induced platelet aggregation in vitro.
Specifications

Chemical Identifiers

894804-07-0
356.382
jnk inhibitor viii, aminopyridine deriv. 2, tcs jnk 6o, n-4-amino-5-cyano-6-ethoxypyridin-2-yl-2-2,5-dimethoxyphenyl acetamide, kinome_3020, d00hva, aminopyridine-based inhibitor 6o, n-4-amino-5-cyano-6-ethoxy-2-pyridinyl-2,5-dimethoxybenzeneacetamide
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
C18H20N4O4
KQMPRSZTUSSXND-UHFFFAOYSA-N
11624601
CCOC1=C(C(=CC(=N1)NC(=O)CC2=C(C=CC(=C2)OC)OC)N)C#N

Specifications

TCS JNK 6o
894804-07-0
jnk inhibitor viii, aminopyridine deriv. 2, tcs jnk 6o, n-4-amino-5-cyano-6-ethoxypyridin-2-yl-2-2,5-dimethoxyphenyl acetamide, kinome_3020, d00hva, aminopyridine-based inhibitor 6o, n-4-amino-5-cyano-6-ethoxy-2-pyridinyl-2,5-dimethoxybenzeneacetamide
CCOC1=C(C(=CC(=N1)NC(=O)CC2=C(C=CC(=C2)OC)OC)N)C#N
356.382
356.38
10mg
C18H20N4O4
KQMPRSZTUSSXND-UHFFFAOYSA-N
N-(4-amino-5-cyano-6-ethoxypyridin-2-yl)-2-(2,5-dimethoxyphenyl)acetamide
11624601
>98%
Videos