missing translation for 'onlineSavingsMsg'
Learn More

TCS JNK 5a, Tocris Bioscience™

Selective inhibitor of JNK2 and JNK3

Brand:  Tocris (Bio-Techne) 282750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13431904

  • £518.91 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective inhibitor of JNK2 and JNK3 (pIC50 values are 6.7, 6.5, <5.0 and <4.8 for JNK3, JNK2, JNK1 and p38α respectively). Displays no significant activity at a range of other protein kinases including EGFR, ErbB2, cdk2, PLK-1 and Src (pIC50 < 5.0).
Specifications

Chemical Identifiers

312917-14-9
332.421
tcs jnk 5a, jnk inhibitor ix, n-3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl naphthalene-1-carboxamide, n-3-cyano-4,5,6,7-tetrahydrobenzo b thiophen-2-yl-1-naphthamide, d0l0hg, tcs jnk 5a jnk inhibitor ix, n-3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl-1-naphthamide, n-3-cyano-4,5,6,7-tetrahydrobenzo b thienyl-2-yl-1-naphthalenecarboxamide, naphthalene-1-carboxylic acid 3-cyano-4,5,6,7-tetrahydro-benzo b thiophen-2-yl-amide
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
C20H16N2OS
WQGDQGAFSDMBLA-UHFFFAOYSA-N
766949
C1CCC2=C(C1)C(=C(S2)NC(=O)C3=CC=CC4=CC=CC=C43)C#N

Specifications

50mg
312917-14-9
tcs jnk 5a, jnk inhibitor ix, n-3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl naphthalene-1-carboxamide, n-3-cyano-4,5,6,7-tetrahydrobenzo b thiophen-2-yl-1-naphthamide, d0l0hg, tcs jnk 5a jnk inhibitor ix, n-3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl-1-naphthamide, n-3-cyano-4,5,6,7-tetrahydrobenzo b thienyl-2-yl-1-naphthalenecarboxamide, naphthalene-1-carboxylic acid 3-cyano-4,5,6,7-tetrahydro-benzo b thiophen-2-yl-amide
C1CCC2=C(C1)C(=C(S2)NC(=O)C3=CC=CC4=CC=CC=C43)C#N
332.421
332.42
TCS JNK 5a
C20H16N2OS
WQGDQGAFSDMBLA-UHFFFAOYSA-N
N-(3-cyano-4,5,6,7-tetrahydro-1-benzothiophen-2-yl)naphthalene-1-carboxamide
766949
>99%
Videos
Documents