missing translation for 'onlineSavingsMsg'
Learn More

TCS 5861528, Tocris Bioscience™

TRPA1 blocker

Brand:  Tocris (Bio-Techne) 393850

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13443844

  • £627.69 / 50mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TRPA1 channel blocker that antagonizes AITC- and 4-HNE-evoked calcium influx (IC50 values are 14.3 and 18.7μM respectively). Attenuates diabetic hypersensitivity in an in vivo rat model.
Specifications

Chemical Identifiers

332117-28-9
369.425
chembridge-5861528, n-4-butan-2-ylphenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, 2-1,3-dimethyl-2,6-dioxopurin-7-yl-n-4-sec-butyl phenyl acetamide, d0j0bv, n-4-2r-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, n-4-sec-butylphenyl-2-1,3-dimethyl-2,6-dioxo-2,3-dihydro-1h-purin-7 6h-yl acetamide, 2-1,3-dimethyl-2,6-dioxo 1,3,7-trihydropurin-7-yl-n-4-methylpropyl phenyl acetamide, 2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl-n-4-1-methylpropyl phenyl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1h-purin-7-yl acetamide
N-(4-butan-2-ylphenyl)-2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetamide
C19H23N5O3
ZUTUWJYMCADJHD-UHFFFAOYSA-N
2873523
CCC(C)C1=CC=C(C=C1)NC(=O)CN2C=NC3=C2C(=O)N(C(=O)N3C)C

Specifications

50mg
332117-28-9
chembridge-5861528, n-4-butan-2-ylphenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, 2-1,3-dimethyl-2,6-dioxopurin-7-yl-n-4-sec-butyl phenyl acetamide, d0j0bv, n-4-2r-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxopurin-7-yl acetamide, n-4-sec-butylphenyl-2-1,3-dimethyl-2,6-dioxo-2,3-dihydro-1h-purin-7 6h-yl acetamide, 2-1,3-dimethyl-2,6-dioxo 1,3,7-trihydropurin-7-yl-n-4-methylpropyl phenyl acetamide, 2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl-n-4-1-methylpropyl phenyl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7h-purin-7-yl acetamide, n-4-butan-2-yl phenyl-2-1,3-dimethyl-2,6-dioxo-2,3,6,7-tetrahydro-1h-purin-7-yl acetamide
CCC(C)C1=CC=C(C=C1)NC(=O)CN2C=NC3=C2C(=O)N(C(=O)N3C)C
369.425
369.42
TCS 5861528
C19H23N5O3
ZUTUWJYMCADJHD-UHFFFAOYSA-N
N-(4-butan-2-ylphenyl)-2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetamide
2873523
>98%
Videos
Documents