missing translation for 'onlineSavingsMsg'
Learn More

TCS 359, Tocris Bioscience™

Potent FLT3 inhibitor

Brand:  Tocris (Bio-Techne) 259150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13441534

  • £425.12 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of FLT3 receptor tyrosine kinase (IC50 = 42 nM) that displays selectivity over a range of other kinases. Exhibits antiproliferative effects on MV4-11 cells, a human acute myelogenous leukemia cell line expressing a constitutively active mutant FLT3 (IC50 = 340 nM).
Specifications

Chemical Identifiers

301305-73-7
360.428
flt-3 inhibitor, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, 2-3,4-dimethoxy-benzoylamino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-benzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide,2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro, acmc-20ekhy, d0x7jk, qcr-73
2-[(3,4-dimethoxybenzoyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
C18H20N2O4S
FSPQCTGGIANIJZ-UHFFFAOYSA-N
1048845
COC1=C(C=C(C=C1)C(=O)NC2=C(C3=C(S2)CCCC3)C(=O)N)OC

Specifications

TCS 359
301305-73-7
flt-3 inhibitor, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydrobenzo b thiophene-3-carboxamide, 2-3,4-dimethoxy-benzoylamino-4,5,6,7-tetrahydro-benzo b thiophene-3-carboxylic acid amide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzamido-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide, 2-3,4-dimethoxybenzoyl amino-4,5,6,7-benzo b thiophene-3-carboxamide, benzo b thiophene-3-carboxamide,2-3,4-dimethoxybenzoyl amino-4,5,6,7-tetrahydro, acmc-20ekhy, d0x7jk, qcr-73
COC1=C(C=C(C=C1)C(=O)NC2=C(C3=C(S2)CCCC3)C(=O)N)OC
360.428
360.43
50mg
C18H20N2O4S
FSPQCTGGIANIJZ-UHFFFAOYSA-N
2-[(3,4-dimethoxybenzoyl)amino]-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide
1048845
>99%
Videos