missing translation for 'onlineSavingsMsg'
Learn More

TCS 2002, Tocris Bioscience™

Potent GSK-3β inhibitor

Brand:  Tocris (Bio-Techne) 386910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433734

  • £158.76 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent inhibitor of GSK-3β (IC50 = 35 nM). Oral adminstration inhibits cold water stress-induced tau hyperphosphorylation in the mouse brain.
Specifications

Chemical Identifiers

1005201-24-0
338.381
2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, mmbo, 1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl, 1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl, 2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazole
2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole
C18H14N2O3S
RCKYSTKYIVULEK-UHFFFAOYSA-N
24855958
CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C

Specifications

10mg
1005201-24-0
2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, mmbo, 1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl, 1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl, 2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazole
CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C
338.381
338.38
TCS 2002
C18H14N2O3S
RCKYSTKYIVULEK-UHFFFAOYSA-N
2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole
24855958
>99%
Videos
Documents