missing translation for 'onlineSavingsMsg'
Learn More

TCS 1205, Tocris Bioscience™

Subtype-selective GABAA receptor agonist

Brand:  Tocris (Bio-Techne) 394110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463854

  • £124.20 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

GABAA α2 agonist and GABAA α1 partial agonist in vitro (Ki values are 14 and 121 nM respectively). Exhibits non-sedative anxiolytic activity in vivo.
Specifications

Chemical Identifiers

355022-97-8
337.335
chembl75456, 5-nitro-?-oxo-n-1r-phenylethyl-1h-indole-3-acetamide, r-2-5-nitro-1h-indol-3-yl-2-oxo-n-1-phenylethyl acetamide, 2-5-nitro-1h-indol-3-yl-2-oxo-n-1-phenyl-ethyl-acetamide
2-(5-nitro-1H-indol-3-yl)-2-oxo-N-[(1R)-1-phenylethyl]acetamide
C18H15N3O4
VCKKJKQZHHPPDR-LLVKDONJSA-N
10315001
CC(C1=CC=CC=C1)NC(=O)C(=O)C2=CNC3=C2C=C(C=C3)[N+](=O)[O-]

Specifications

TCS 1205
355022-97-8
chembl75456, 5-nitro-?-oxo-n-1r-phenylethyl-1h-indole-3-acetamide, r-2-5-nitro-1h-indol-3-yl-2-oxo-n-1-phenylethyl acetamide, 2-5-nitro-1h-indol-3-yl-2-oxo-n-1-phenyl-ethyl-acetamide
CC(C1=CC=CC=C1)NC(=O)C(=O)C2=CNC3=C2C=C(C=C3)[N+](=O)[O-]
337.335
337.33
10mg
C18H15N3O4
VCKKJKQZHHPPDR-LLVKDONJSA-N
2-(5-nitro-1H-indol-3-yl)-2-oxo-N-[(1R)-1-phenylethyl]acetamide
10315001
>99%
Videos