missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCR βF1 Mouse anti-Human, APC, Clone: 8A3 (βF1), BD Pharmingen™
Mouse Monoclonal Antibody
Brand: BD Biosciences 566052
Additional Details : Weight : 0.01000kg
Description
TCR βF1 Monoclonal antibody specifically detects TCR βF1 in Human samples. It is validated for Flow cytometry (Intracellular Staining)) .Specifications
TCR βF1 | |
Monoclonal | |
APC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry, Flow Cytometry (Intracellular Staining) | |
8A3 (βF1) | |
Vol. Per test: 5μL | |
TCRb; TCRB; TCR beta; TCR-beta; TRB@; TCR Beta F1; TCR BF1; TCR Cβ | |
Purified Human αβ TCR Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions and unconjugated antibody and free APC were removed. | |
IgG1 κ |
For Reseach Use Only.