Learn More
TCR beta Monoclonal Antibody (H57-597), PerCP-Cyanine5.5, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£117.00 - £249.00
Specifications
Antigen | TCR beta |
---|---|
Clone | H57-597 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15361200
|
Affymetrix eBioscience
45-5961-80 |
25 μg |
£117.00
25µg |
Estimated Shipment: 09-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15371200
|
Affymetrix eBioscience
45-5961-82 |
100 μg |
£249.00
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells. Applications Reported: This H57-597 antibody has been reported for use in flow cytometric analysis. Applications Tested: This H57-597 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 695 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
TCR beta Monoclonal antibody specifically detects TCR beta in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR beta | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
bTCR; FLJ22602; MGC117436; MGC22624; MGC23964; MGC71411; T cell receptor beta chain; t-cell antigen receptor; T-cell receptor; TCR beta; Tcra; Tcrb; TCRbeta; TCRD; Tib; TRA | |
Tcrb | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Tcrb |
H57-597 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
Armenian Hamster | |
Mouse | |
0 | |
21577 | |
IgG | |
Affinity chromatography | |
Antibody |
For Research Use Only.