TCR beta Armenian Hamster anti-Mouse, PerCP-Cy5.5, Clone: H57-597, eBioscience™

Armenian Hamster Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 45-5961-82

100UG ANTI-MOUSE TCR BETA PERCP-CY5.5

UNSPSC: 12352200

Code: Z2

Additional Details:
Additional Details: Weight: 0.25000kg



Disclaimers: For Research Use Only.

Product Code. 15371200

Quantity Price
1 £ 186.31 / 100µg
EU Stock 5
Add to basket


Description and Specification

Specification

Antigen TCR beta
Format Conjugated
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Host Species Armenian Hamster
Applications Flow Cytometry
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Species Reactivity Mouse
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone H57-597
Concentration 0.2mg/mL
Conjugate PerCP-Cy5.5

The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells.