missing translation for 'onlineSavingsMsg'
Learn More

TCR beta, Biotin, clone: H57-597, eBioscience™

Armenian Hamster Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-5961-85

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15229959

  • £146.13 / 500µg


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

Description: The H57-597 monoclonal antibody reacts with the beta chain of mouse TCR. TCR beta is expressed by thymocytes in a developmentally regulated manner and a majority of peripheral T cells. Crosslinking of the TCR complex with H57-597 induces activation and proliferation of T cells or apoptosis based on assay conditions. H57-597 is used as a phenotypic marker for TCR beta expressing T cells.Applications Reported: The H57-597 antibody has been reported for use in flow cytometric analysis.Applications Tested: The H57-597 antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

TCR beta
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Tcrb
IgG
500μg
Store at 2-8°C. Do not freeze.
Primary
21577
Flow Cytometry
H57-597
Biotin
Tcrb
TCRbeta, Tib
Armenian Hamster
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.