missing translation for 'onlineSavingsMsg'
Learn More

TCN 213, Tocris Bioscience™

NMDA antagonist; selective for NR2A over NR2B

Brand:  Tocris (Bio-Techne) 416350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13464264

  • £671.97 / 50mg


Estimated Shipment
08-01-2019
Add to basket

Description

Description

NMDA receptor antagonist, selective for NR1/NR2A (pIC50 = 5.4) over NR1/NR2B.
Specifications

Chemical Identifiers

556803-08-8
376.537
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
C18H24N4OS2
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2608149
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3

Specifications

TCN 213
556803-08-8
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3
376.537
376.54
50mg
C18H24N4OS2
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
2608149
>99%
Videos