missing translation for 'onlineSavingsMsg'
Learn More

TCN 213, Tocris Bioscience™

NMDA antagonist; selective for GluN2A over GluN2B containing receptors

Brand:  Tocris (Bio-Techne) 416310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13454264

  • £147.00 / 10mg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NMDA receptor antagonist, selective for GluN1/GluN2A (formally NR1/NR2A) (pIC50 = 5.4) over GluN1/GluN2B (formally NR1/NR2B) containing receptors.

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Specifications

Chemical Identifiers

556803-08-8
376.537
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
C18H24N4OS2
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2608149
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3

Specifications

10mg
556803-08-8
2-5-benzylamino-1,3,4-thiadiazol-2-yl sulfanyl-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-yl thio-n-cyclohexylmethyl acetamide, 2-5-benzylamino-1,3,4-thiadiazol-2-ylthio-n-cyclohexylmethyl acetamide, n-cyclohexylmethyl-2-5-phenylmethyl amino-1,3,4-thiadiazol-2-yl thio acetamide
C1CCC(CC1)CNC(=O)CSC2=NN=C(S2)NCC3=CC=CC=C3
376.537
376.54
TCN 213
C18H24N4OS2
XBAZPYFIYYCZBO-UHFFFAOYSA-N
2-[[5-(benzylamino)-1,3,4-thiadiazol-2-yl]sulfanyl]-N-(cyclohexylmethyl)acetamide
2608149
>99%
Videos
Documents