missing translation for 'onlineSavingsMsg'
Learn More

TCN 201, Tocris Bioscience™

GluN1/GluN2A-selective NMDA antagonist

Brand:  Tocris (Bio-Techne) 415410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13444254

  • £188.76 / 10mg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NMDA receptor antagonist selective for GluN1/GluN2A (formally NR1/NR2A) over GluN1/GluN2B (formally NR1/NR2B) containing receptors (pIC50 values are 6.8 and <4.3, respectively, in human recombinant GluN1/GluN2A (formally NR1/NR2A) and GluN1/GluN2B (formally NR1/NR2B) FLIPR/Ca2+ assays).

Please refer to IUPHAR Guide to Pharmacology for the most recent naming conventions.

Specifications

Chemical Identifiers

852918-02-6
461.892
qcr-22, n-4-2-benzoylhydrazinecarbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide, 3-chloro-4-fluoro-n-4-2-phenylcarbonyl hydrazino carbonyl benzyl benzenesulfonamide, n-4-benzamidocarbamoyl phenyl methyl-3-chloro-4-fluorobenzenesulfonamide, d09hxx, c21h17clfn3o4so6, tcn hplc, 3-chloro-4-fluoro-n-4-phenylformohydrazido carbonyl phenyl methyl benzene-1-sulfonamide, 4-3-chloro-4-fluorophenyl sulfonyl amino methyl benzoic acid 2-benzoylhydrazide, n-4-2-benzoylhydrazine-1-carbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide
N-[[4-(benzamidocarbamoyl)phenyl]methyl]-3-chloro-4-fluorobenzenesulfonamide
C21H17ClFN3O4S
FYIBXBFDXNPBSF-UHFFFAOYSA-N
4787937
C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=C(C=C2)CNS(=O)(=O)C3=CC(=C(C=C3)F)Cl

Specifications

TCN 201
852918-02-6
qcr-22, n-4-2-benzoylhydrazinecarbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide, 3-chloro-4-fluoro-n-4-2-phenylcarbonyl hydrazino carbonyl benzyl benzenesulfonamide, n-4-benzamidocarbamoyl phenyl methyl-3-chloro-4-fluorobenzenesulfonamide, d09hxx, c21h17clfn3o4so6, tcn hplc, 3-chloro-4-fluoro-n-4-phenylformohydrazido carbonyl phenyl methyl benzene-1-sulfonamide, 4-3-chloro-4-fluorophenyl sulfonyl amino methyl benzoic acid 2-benzoylhydrazide, n-4-2-benzoylhydrazine-1-carbonyl benzyl-3-chloro-4-fluorobenzenesulfonamide
C1=CC=C(C=C1)C(=O)NNC(=O)C2=CC=C(C=C2)CNS(=O)(=O)C3=CC(=C(C=C3)F)Cl
461.892
461.89
10mg
C21H17ClFN3O4S
FYIBXBFDXNPBSF-UHFFFAOYSA-N
N-[[4-(benzamidocarbamoyl)phenyl]methyl]-3-chloro-4-fluorobenzenesulfonamide
4787937
>99%
Videos