missing translation for 'onlineSavingsMsg'
Learn More

TCLP Standard 1 For Environmental Analysis, SPEX CertiPrep™

Catalog Number p-220019
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O
This item is not returnable. View return policy
7697-37-2
HNO3
63.01
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
CHEBI:48107
nitric acid
O[N+]([O-])=O

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ba500μg/mL
Ag25μg/mL
As25μg/mL
Cr25μg/mL
Pb25μg/mL
Cd5μg/mL
Se5μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Grade Environmental
Concentration or Composition (by Analyte or Components) Components in 5% HNO3:
As    •25μg/mL
Ba    •500μg/mL
Cd    •5μg/mL
Cr    •25μg/mL
Pb    •25μg/mL
Hg†    •100μg/mL
Se    •5μg/mL
Ag    •25μg/mL
†Supplied separately.
Packaging Triple-leached LDPE bottle
Solvent or Matrix 2% HNO3
Quantity 125 mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Chemical Name or Material TCLP Standard 1

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'