missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553031
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: LSDVEDLQEV EMLECDGLKM DSNKEFVTSN ESTEESSNCE NGSPQKGRGG LGKRRKAPTK KSPLSGVSQE GKQV Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
The basic helix-loop-helix transcription factor TCF21/Pod1 (also called capsulin or epicardin) is involved in kidney, lung and spleen organogenesis. It is also essential for normal development of the testes and ovaries. TCF21/Pod1 is involved in the transcriptional repression of steroidogenic factor 1 (Sf1/Nr5a1/Ad4BP), an orphan nuclear receptor that regulates the expression of multiple genes (including Scc) that mediate sexual differentiation.Specifications
TCF21 | |
Polyclonal | |
Unconjugated | |
TCF21 | |
TCF21; BHLHA23; POD1; Transcription factor 21; Capsulin; Class A basic helix-loop-helix protein 23; Epicardin; Podocyte-expressed 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6943 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43680 | |
TCF21 | |
Recombit protein corresponding to Human TCF21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only