Learn More
Tocris Bioscience™ TCB-2
High affinity and potent 5-HT2A agonist
Brand:  Tocris Bioscience™ 259210
Code : Z1
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 13451534
  • £208.74 / 10mg
Estimated Shipment date
from Supplier 28-02-2020
Description
Description
High affinity 5-HT2A receptor agonist (Ki values are 0.73 and 0.75 nM for rat and human receptors respectively). Potently stimulates IP3 accumulation in NIH3T3 cells stably expressing rat 5-HT2A receptors (EC50 = 36 nM). Induces head twitches and hypothermia in mice following i.p. administration.
Specifications
Specifications
(4-Bromo-3,6-dimethoxybenzocyclobuten-1-yl)methylamine hydrobromide
>99%
Observed MW: 353.05
Soluble to 100mM in DMSO
912342-28-0
C11H14BrNO2.HBr