missing translation for 'onlineSavingsMsg'
Learn More

TC-O 9311, Tocris Bioscience™

Potent and selective GPR139 agonist

Brand:  Tocris (Bio-Techne) 425550

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13464444

  • £639.97 / 50mg


Estimated Shipment
05-04-2019
Add to basket

Description

Description

Potent GPR139 agonist (EC 50 = 39 nM in CHO-K1 cells expressing human GPR139). Displays no activity against a range of 90 diverse targets.
Specifications

Chemical Identifiers

444932-31-4
365.389
2-3,5-dimethoxybenzoyl-n-1-naphthyl hydrazinecarboxamide, 1-3,5-dimethoxybenzoyl amino-3-naphthalen-1-ylurea, n'-1-naphthyl amino carbonyl-3,5-dimethoxybenzhydrazide, 2-3,5-dimethoxyphenyl carbonyl-n-naphthalen-1-yl hydrazinecarboxamide, 3,5-dimethoxybenzoic acid 2-1-naphthalenylamino carbonyl hydrazide
1-[(3,5-dimethoxybenzoyl)amino]-3-naphthalen-1-ylurea
C20H19N3O4
KPTMSQHTGZMEFU-UHFFFAOYSA-N
3376937
COC1=CC(=CC(=C1)C(=O)NNC(=O)NC2=CC=CC3=CC=CC=C32)OC

Specifications

TC-O 9311
444932-31-4
2-3,5-dimethoxybenzoyl-n-1-naphthyl hydrazinecarboxamide, 1-3,5-dimethoxybenzoyl amino-3-naphthalen-1-ylurea, n'-1-naphthyl amino carbonyl-3,5-dimethoxybenzhydrazide, 2-3,5-dimethoxyphenyl carbonyl-n-naphthalen-1-yl hydrazinecarboxamide, 3,5-dimethoxybenzoic acid 2-1-naphthalenylamino carbonyl hydrazide
COC1=CC(=CC(=C1)C(=O)NNC(=O)NC2=CC=CC3=CC=CC=C32)OC
365.389
365.38
50mg
C20H19N3O4
KPTMSQHTGZMEFU-UHFFFAOYSA-N
1-[(3,5-dimethoxybenzoyl)amino]-3-naphthalen-1-ylurea
3376937
>98%
Videos