Learn More
TC-H 106, Tocris Bioscience™
Class I histone deacetylase inhibitor
Brand:  Tocris (Bio-Techne) 427050
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13464464
  • £710.01 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Class I histone deacetylase (HDAC) inhibitor (IC50 values are 150, 370, 760 and 5000 nM for HDAC1, HDAC3, HDAC2 and HDAC8 respectively). Exhibits slow, tight-binding inhibitory activity. Displays no activity against class II HDACs. Brain penetrant.
Specifications
Chemical Identifiers
937039-45-7
339.439
pimelic diphenylamide 106, n'-2-aminophenyl-n-4-methylphenyl heptanediamide, n1-2-aminophenyl-n7-4-methylphenyl-heptanediamide, tc-h 106;pimelic diphenylamide 106, n1-2-aminophenyl-n7-p-tolylheptanediamide, pimelic diphenylamide hplc, n1-2-aminophenyl-n7-p-tolyl heptanediamide, n-4-methylphenyl-n'-2-aminophenyl heptanediamide, n1-2-aminophenyl-n7-4-methylphenyl heptanediamide, n~1~-2-aminophenyl-n~7~-4-methylphenyl heptanediamide
N'-(2-aminophenyl)-N-(4-methylphenyl)heptanediamide
C20H25N3O2
WTKBRPXPNAKVEQ-UHFFFAOYSA-N
16070100
CC1=CC=C(C=C1)NC(=O)CCCCCC(=O)NC2=CC=CC=C2N
Specifications
50mg
937039-45-7
pimelic diphenylamide 106, n'-2-aminophenyl-n-4-methylphenyl heptanediamide, n1-2-aminophenyl-n7-4-methylphenyl-heptanediamide, tc-h 106;pimelic diphenylamide 106, n1-2-aminophenyl-n7-p-tolylheptanediamide, pimelic diphenylamide hplc, n1-2-aminophenyl-n7-p-tolyl heptanediamide, n-4-methylphenyl-n'-2-aminophenyl heptanediamide, n1-2-aminophenyl-n7-4-methylphenyl heptanediamide, n~1~-2-aminophenyl-n~7~-4-methylphenyl heptanediamide
CC1=CC=C(C=C1)NC(=O)CCCCCC(=O)NC2=CC=CC=C2N
339.439
339.43
TC-H 106
C20H25N3O2
WTKBRPXPNAKVEQ-UHFFFAOYSA-N
N'-(2-aminophenyl)-N-(4-methylphenyl)heptanediamide
16070100
>99%