missing translation for 'onlineSavingsMsg'
Learn More

TC-F 2, Tocris Bioscience™

Potent, reversible and selective FAAH inhibitor

Brand:  RD Systems (Bio-Techne) 4355/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464614

  • £153.12 / 10mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Potent, reversible inhibitor of fatty acid amide hydrolase (FAAH) (IC 50 values are 28 and 100 nM for human and rat FAAH respectively). Noncovalent inhibitor; displays selectivity for FAAH over cannabinoid-related targets (IC 50 > 20 μM for CB 1 CB 2 and TRPV1). Active in vivo.
Specifications

Chemical Identifiers

1304778-15-1
439.519
tc-f 2, 1-3s-1-4-2-benzofuranyl-2-pyrimidinyl-3-piperidinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-3s-1-4-1-benzofuran-2-yl pyrimidin-2-yl piperidin-3-yl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-ethyl-3-1-4-benzofuran-2-yl pyrimidine-2-yl piperidine-3beta-yl-1h-benzoimidazole-2 3h-one, qj9
1-[(3S)-1-[4-(1-benzofuran-2-yl)pyrimidin-2-yl]piperidin-3-yl]-3-ethylbenzimidazol-2-one
C26H25N5O2
NXTBLPPTZRPJCA-IBGZPJMESA-N
25198728
CCN1C2=CC=CC=C2N(C1=O)C3CCCN(C3)C4=NC=CC(=N4)C5=CC6=CC=CC=C6O5

Specifications

TC-F 2
1304778-15-1
tc-f 2, 1-3s-1-4-2-benzofuranyl-2-pyrimidinyl-3-piperidinyl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-3s-1-4-1-benzofuran-2-yl pyrimidin-2-yl piperidin-3-yl-3-ethyl-1,3-dihydro-2h-benzimidazol-2-one, 1-ethyl-3-1-4-benzofuran-2-yl pyrimidine-2-yl piperidine-3beta-yl-1h-benzoimidazole-2 3h-one, qj9
CCN1C2=CC=CC=C2N(C1=O)C3CCCN(C3)C4=NC=CC(=N4)C5=CC6=CC=CC=C6O5
439.519
439.51
10mg
C26H25N5O2
NXTBLPPTZRPJCA-IBGZPJMESA-N
1-[(3S)-1-[4-(1-benzofuran-2-yl)pyrimidin-2-yl]piperidin-3-yl]-3-ethylbenzimidazol-2-one
25198728
>99%
Videos