missing translation for 'onlineSavingsMsg'
Learn More

TC-DAPK 6, Tocris Bioscience™

Potent and selective inhibitor of DAPK1

Brand:  RD Systems (Bio-Techne) 4301/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13444524

  • £438.06 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective, ATP-competitive inhibitor of death-associated protein kinase 1 (DAPK1) (IC50 values are 69 and 225 nM for DAPK1 and DAPK3 respectively, when assayed with 10 μM ATP). Displays selectivity for DAPK1 over a range of 48 other kinases, including Abl, AMPK, Chk1, Met and Src (IC50 > 10 μM).
Specifications

Chemical Identifiers

315694-89-4
276.295
tc-dapk 6, 4e-2-e-2-phenylethenyl-4-pyridin-3-ylmethylidene-1,3-oxazol-5-one, tc-dapk-6, 2-2-phenylethenyl-4-pyridin-3-ylmethylidene-4,5-dihydro-1,3-oxazol-5-one, 2-2-phenylethenyl-4-pyridin-3-yl methylidene-4,5-dihydro-1,3-oxazol-5-one, 2-styryl-4-3-pyridylmethylene-2-oxazoline-5-one, e-4-pyridin-3-ylmethylene-2-e-styryl oxazol-5 4h-one
(4E)-2-[(E)-2-phenylethenyl]-4-(pyridin-3-ylmethylidene)-1,3-oxazol-5-one
C17H12N2O2
GFGMISOSPOPSHN-QCTDOKRBSA-N
5676111
C1=CC=C(C=C1)C=CC2=NC(=CC3=CN=CC=C3)C(=O)O2

Specifications

50mg
315694-89-4
tc-dapk 6, 4e-2-e-2-phenylethenyl-4-pyridin-3-ylmethylidene-1,3-oxazol-5-one, tc-dapk-6, 2-2-phenylethenyl-4-pyridin-3-ylmethylidene-4,5-dihydro-1,3-oxazol-5-one, 2-2-phenylethenyl-4-pyridin-3-yl methylidene-4,5-dihydro-1,3-oxazol-5-one, 2-styryl-4-3-pyridylmethylene-2-oxazoline-5-one, e-4-pyridin-3-ylmethylene-2-e-styryl oxazol-5 4h-one
C1=CC=C(C=C1)C=CC2=NC(=CC3=CN=CC=C3)C(=O)O2
276.295
276.29
TC-DAPK 6
C17H12N2O2
GFGMISOSPOPSHN-QCTDOKRBSA-N
(4E)-2-[(E)-2-phenylethenyl]-4-(pyridin-3-ylmethylidene)-1,3-oxazol-5-one
5676111
>98%
Videos
Documents