Learn More
TBB, Tocris Bioscience™
Selective cell-permeable CK2 inhibitor
Brand:  Tocris (Bio-Techne) 227510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13460914
  • £106.56 / 10mg
Estimated Shipment
14-09-2020
Description
Description
A cell-permeable, selective inhibitor of casein kinase-2 (CK2) (IC50 = 0.9 and 1.6 μM for rat liver and human recombinant CK2 respectively). Exhibits modest discrimination between CK2 subunits, with Ki values ranging from 80 nM to 210 nM. Acts in an ATP/GTP-competitive manner and displays one to two orders of magnitude selectivity over a panel of 33 protein kinases.
Specifications
Chemical Identifiers
17374-26-4
434.711
4,5,6,7-tetrabromobenzotriazole, tbb, tbbt, 4,5,6,7-tetrabromo-1h-benzotriazole, 4,5,6,7-tetrabromo-2-azabenzimidazole, ck2 inhibitor, casein kinase ii inhibitor i, 4,5,6,7-tetrabromo-1h-benzo d 1,2,3 triazole, tetrabromo-2-benzotriazole, 4,5,6,7-tetrabromo-1h-1,2,3-benzotriazole
4,5,6,7-tetrabromo-2H-benzotriazole
C6HBr4N3
OMZYUVOATZSGJY-UHFFFAOYSA-N
1694
C1(=C(C2=NNN=C2C(=C1Br)Br)Br)Br
Specifications
TBB
17374-26-4
4,5,6,7-tetrabromobenzotriazole, tbb, tbbt, 4,5,6,7-tetrabromo-1h-benzotriazole, 4,5,6,7-tetrabromo-2-azabenzimidazole, ck2 inhibitor, casein kinase ii inhibitor i, 4,5,6,7-tetrabromo-1h-benzo d 1,2,3 triazole, tetrabromo-2-benzotriazole, 4,5,6,7-tetrabromo-1h-1,2,3-benzotriazole
C1(=C(C2=NNN=C2C(=C1Br)Br)Br)Br
434.711
434.71
10mg
C6HBr4N3
OMZYUVOATZSGJY-UHFFFAOYSA-N
4,5,6,7-tetrabromo-2H-benzotriazole
1694
>99%