missing translation for 'onlineSavingsMsg'
Learn More

Taurodeoxycholic acid, sodium salt hydrate, 97%, ACROS Organics™

Brand:  Acros Organics 336840250

Code : 29

Additional Details : CAS Number : 207737-97-1 Weight : 0.02500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10530034

  • £455.00 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

Taurodeoxycholic acid, sodium salt hydrate
207737-97-1
97%
521.69
Sodium taurodeoxylate
97%
≥96.0%, 1.0% max. taurine, 1.0% max. sodium taurocholate, 1.0% max. sodium deocycholate (TLC)
C26H44NNaO6S·xH2O
MFCD00149238

Specifications

Taurodeoxycholic acid, sodium salt hydrate
207737-97-1
0.5% max.
Crystalline Powder
97%
Authentic
MFCD00149238
Sodium taurodeoxylate
7% Max. (K.F.)
White to off-white
97%
(5% aq. soln.) clear colorless to faint yellow
20ppm max.
≥96.0%, 1.0% max. taurine, 1.0% max. sodium taurocholate, 1.0% max. sodium deocycholate (TLC)
C26H44NNaO6S·xH2O
521.69
Glass bottle
+34.5° to +36.5° (20°C, 589nm) (c=2.5, water) (calculated on dry substance)
+33.00 (20.00°C c=3,water)
25g
Videos
SDS