Learn More
Taurodeoxycholic acid, sodium salt hydrate, 97%, ACROS Organics™
Brand: Acros Organics 336840050
Code : 29
Additional Details : CAS Number : 1180-95-6 Weight : 0.05500kg
Packaging | Glass bottle |
---|---|
Quantity | 5g |
Chemical Identifiers
207737-97-1 | |
539.704 | |
OLPIZAYYAVQETM-GGPRKOIFSA-M | |
23702150 | |
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C.O.[Na+] |
C26H46NNaO7S | |
MFCD00149238 | |
Sodium taurodeoxylate | |
sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate;hydrate |
Specifications
Taurodeoxycholic acid, sodium salt hydrate | |
207737-97-1,207737-97-1 | |
0.5% max. | |
20ppm max. | |
MFCD00149238 | |
Sodium taurodeoxylate | |
OLPIZAYYAVQETM-GGPRKOIFSA-M | |
sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate;hydrate | |
23702150 | |
Crystalline Powder | |
+34.5° to +36.5° (20°C, 589nm) (c=2.5, water) (calculated on dry substance) | |
+33.00 (20.00°C c=3,water) | |
5g |
97% | |
(5% aq. soln.) clear colorless to faint yellow | |
97% | |
C26H46NNaO7S | |
Authentic | |
Glass bottle | |
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C.O.[Na+] | |
539.704 | |
521.69 | |
≥96.0%, 1.0% max. taurine, 1.0% max. sodium taurocholate, 1.0% max. sodium deocycholate (TLC) | |
7% Max. (K.F.) | |
White |
Safety and Handling
- Taurodeoxycholic acid, sodium salt hydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.