missing translation for 'onlineSavingsMsg'
Learn More

Taurocholic acid, sodium salt hydrate, 98%, ACROS Organics™

Brand:  Acros Organics 230370250

Code : 29

Additional Details : CAS Number : 345909-26-4 Weight : 0.07500kg

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 10629453

  • £259.20 / 25g


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Chemical Identifiers

Taurocholic acid, sodium salt hydrate
98%
537.70
≥97.5% (TLC)
C26H44NNaO7S·xH2O
Sodium taurocholate hydrate

Specifications

Taurocholic acid, sodium salt hydrate
20ppm max.
≥97.5% (TLC)
C26H44NNaO7S·xH2O
537.70
Sodium taurocholate hydrate
+20.0° to +25.8° (20°C, 589nm) (c=3, water)
+23.00 (20.00°C c=3 H2O)
25g
345909-26-4
Powder
98%
Authentic
Glass bottle
11.0% to 14.0%
7% max.
White to cream-white
Videos
SDS