missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taurocholic acid sodium salt, ≥97%, Practical grade, MP Biomedicals™
Click to view available options
Quantity:
25 g
Specifications
Color | Brown-Yellow |
Physical Form | Powder |
Quantity | 25 g |
Formula Weight | 537.7 |
Percent Purity | ≥97% |
Grade | Practical |
Chemical Name or Material | Taurocholic acid sodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'