missing translation for 'onlineSavingsMsg'
Learn More

Tau Ab-2 Mouse Monoclonal Antibody, Epredia™

Provide accurate, reproducible results with the Epredia™ Tau Ab-2, Mouse Monoclonal Antibody.

£114.00 - £617.10

Specifications

Antigen TAU Ab-2
Clone TAU-5
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Form Concentrated
View More Specs

Products 8
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
Product Code Brand Conjugate Concentration Quantity Price Quantity & Availability  
12650677
View Documents Promotion Details
Epredia
MS-247-P
Unconjugated 200μg/mL 1 mL
£520.00
1mL
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12680677
View Documents Promotion Details
Epredia
MS-247-P1ABX
Unconjugated 1mg/mL 100 μL
£370.00
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 20-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
12620677
View Documents Promotion Details
Epredia
MS-247-B
Biotin 200μg/mL 1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12660677
View Documents Promotion Details
Epredia
MS-247-P0
Unconjugated 200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12670677
View Documents Promotion Details
Epredia
MS-247-P1
Unconjugated 200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12630677
View Documents Promotion Details
Epredia
MS-247-B0
Biotin 200μg/mL 100 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12640677
View Documents Promotion Details
Epredia
MS-247-B1
Biotin 200μg/mL 500 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12690677
View Documents Promotion Details
Epredia
MS-247-PABX
Unconjugated 1mg/mL 200 μL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Tau proteins are microtubule-associated proteins (MAPs). Alternate splicing of tau mRNA and differential phosphorylation contribute to the heterogeneity of tau.

Host Species: Mouse

Clone: TAU-5

Isotype: IgG1

Species Reactivity: Human, Sheep, Cow, Mouse and Rat. Others not known.

Epitope: Middle of Tau

Immunogen: Purified bovine microtubule-associated proteins

Molecular Weight: 45-68kDa

Positive Control: IMR5 cells, Human T98G glioblastoma cells or Alzheimer's disease brain tissue

Cellular Localization: Cytoplasmic Ab-2 reacts with the non-phosphorylated as well as the phosphorylated forms of tau and its epitope maps in the middle of tau. Ab-2 intensely stains the human neurofibrillary tangles, neuropil threads, and neuritic plaques associated with Alzheimer's disease. Ab-2 shows no cross reaction with other MAPs or tubulin.

Recommended for:

  • Immunohistochemistry (Not suitable)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

TAU Ab-2
Immunofluorescence, Immunoprecipitation, Western Blot
Concentrated
RUO
Bovine, Human, Mouse, Rat, Sheep
IgG1
TAU-5
Monoclonal
Mouse
Neuroscience
Purified bovine microtubule-associated proteins
Primary
Videos
SDS
Documents

Documents

Product Certifications