missing translation for 'onlineSavingsMsg'
Learn More

Tashiro’s indicator solution, In ethanol, Honeywell Fluka™

Brand:  Honeywell Fluka 36083-1L

Code : NEW

Additional Details : Weight : 0.79000kg Transport : UN number : 1170 Chem class : 3 Pack group : II

Quantity 1L

 View more versions of this product

Product Code. 15635110

  • £46.15 / 1 L


Estimated Shipment date
from Supplier 31-07-2018
Add to basket

Specifications

Chemical Identifiers

Tashiro’s indicator solution
12°C (53.6°F)
MFCD00148563
UN1170

Specifications

Tashiro’s indicator solution
C2H6O
UN1170
CCO
46.069
CHEBI:16236
12°C (53.6°F)
64-17-5
MFCD00148563
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
1L
Videos