missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R38 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567748
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse and Rat samples.
TAS2R38 Polyclonal antibody specifically detects TAS2R38 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
TAS2R38 | |
Polyclonal | |
Unconjugated | |
TAS2R38 | |
mt2r31; PTC; PTC bitter taste receptor; RGD:1560649}; T2R138; T2R26; T2r31; T2r38; T2r38 {ECO:0000312; T2R61; Tas2r138; Tas2r26; TAS2R38; taste 2 receptor member 38; taste receptor type 2 member 26; Taste receptor type 2 member 38; taste receptor type 2 member 61; taste receptor, type 2, member 138; taste receptor, type 2, member 38 | |
Rabbit | |
affinity chromatography | |
RUO | |
387513, 500091, 5726 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P59533, Q4VHE7, Q7TQA6 | |
Tas2r138, TAS2R38 | |
Synthesized peptide derived from human TAS2R38. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |