missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ OpenArray™ Real-Time PCR Master Mix
TaqMan™ OpenArray™ Real-Time PCR Master Mix is optimized for use with TaqMan™ OpenArray™ gene expression panels, and yields reproducible, specific, and sensitive results.
£158.00
Specifications
Content And Storage | Store at -5 to -30°C until first use, then at 2–8°C. |
---|---|
Detection Method | Primer-probe |
Form | Liquid |
Format | Tube |
PCR Method | 2-step RT-qPCR |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15591114
|
Applied Biosystems™
4462159 |
1.5 mL |
£158.00
1.50mL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Hot start chemistry to provide room temperature stability
- Optimized for use with TaqMan™ OpenArray™ gene expression panels yielding reproducible, specific, and sensitive results
- For use with OpenArray™ MicroRNA panels, OpenArray™ gene expression pathway panels, and other gene expression applications on the OpenArray™ format
- Ready for use with no additional additives required
Order Info
Shipping Conditions: Dry Ice
Specifications
Store at -5 to -30°C until first use, then at 2–8°C. | |
Liquid | |
2-step RT-qPCR | |
Fluorescence Intensity | |
OpenArray™, TaqMan™ | |
Target-Specific | |
Built-In Hot Start |
Primer-probe | |
Tube | |
Standard | |
OpenArray™ | |
Dry Ice | |
miRNA analysis | |
Total RNA |
For Research Use Only. Not for use in diagnostic procedures.