missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Tantalum(V) chloride is used to form mixed chloro-aryloxide compounds, such as [Ta(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 242°C |
Melting Point | 216°C |
Physical Form | Resublimed Crystalline |
Quantity | 100 g |
Assay Percent Range | 99.99%, metal basis |
Solubility Information | Soluble in water (reacts). Also soluble in ethanol,ether and carbon tetrachloride. |
Formula Weight | 358.21 |
Sensitivity | Moisture sensitive |
Density | 3.68 |
Chemical Name or Material | Tantalum(V) Chloride, Ampouled under argon |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'