missing translation for 'onlineSavingsMsg'
Learn More

Tantalum rod, 4.5mm (0.18 in.) dia., Puratronic™, 99.96% (metals basis), Nb nominal 50ppm, Thermo Scientific Chemicals

Catalog Number 15454745
Click to view available options
Quantity:
100 mm
5 x 100 mm
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]
This item is not returnable. View return policy
7440-25-7
Ta
180.95
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
23956
CHEBI:33348
tantalum
[Ta]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 100 mm
Percent Purity 99.96%
Grade Puratronic™
Odor Odorless
Weight ≈2.64g/cm
Form Rod
Assay (metals basis)
Chemical Name or Material Tantalum rod, 4.5mm (0.18 in.) dia., Nb nominal 50ppm

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'