missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tannic acid, ACS, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Tannic acid has anti-bacterial, anti-enzymatic and astringent applications. Used in the dyeing process for cellulose fibers such as cotton. Used in food and medication.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∽218°C (decomposition) |
Beilstein | 8186396 |
Solubility Information | Soluble in water,warm glycerol,alcohol and acetone. |
Formula Weight | 1701.23 |
Grade | ACS Reagent |
Quantity | 500 g |
Flash Point | 199°C (390°F) |
Physical Form | Powder |
Sensitivity | Air and light sensitive |
Chemical Name or Material | Tannic Acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'