missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Tankyrase 1 Colorimetric Activity Assay, R&D Systems™

a high-throughput, semi-quantitative ELISA designed to measure Tankyrase 1 activity.

Brand:  R&D Systems™ 4700-096-K

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13465044

  • £1173.70 / 96 tests


Estimated Shipment
29-03-2019
Add to basket

Includes:
  • Histone-coated 96-stripwell Microplate
  • Anti-PAR Monoclonal Antibody
  • HRP-conjugated Goat Anti-Mouse Antibody
  • Tankyrase 1
  • XAV939a
  • Diluents & Buffers
  • Unmodified NAD Substrate
  • HRP Substrat

Description

Description

Features
  • Amenable to high-throughput
  • Highly sensitive: capable of measuring 0.1 mUnits of Tankyrase 1
  • 3 hour assay time
  • Provides full length Tankyrase 1 enzyme and unmodified NAD substrate
Specifications

Specifications

HRP
ELISA
1 kit
Multi-species
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA kit
0.1mU
96 Tests
Videos