missing translation for 'onlineSavingsMsg'
Learn More

Tandospirone hydrochloride, Tocris Bioscience™

Selective 5-HT1A partial agonist

Brand:  Tocris (Bio-Techne) 285450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461944

  • £516.12 / 50mg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

5-HT1A receptor partial agonist (Ki = 27 nM) that displays selectivity over 5-HT2, 5-HT1C, α1, α2, D1 and D2 receptors (Ki values ranging from 1300 - 41000 nM). Inactive at 5-HT uptake sites, 5-HT1B, β-adrenergic, muscarinic and benzodiazepine receptors. Displays anxiolytic activity.
Specifications

Chemical Identifiers

99095-10-0
419.954
tandospirone hydrochloride, unii-rwr6slb9p6, rwr6slb9p6, metanopirone, sediel, tandospirone hydrochloride mi, 4,7-methano-1h-isoindole-1,3 2h-dione, hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-, hydrochloride 1:1 , 3ar,4s,7r,7as-rel, 3ar,4s,7r,7as-rel-hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-4,7-methano-1h-isoindole-1,3 2h-dione hydrochloride, 4,7-methano-1h-isoindole-1,3 2h-dione, hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-, hydrochloride, 3ar,4s,7r,7as-rel
C1CC2CC1C3C2C(=O)N(C3=O)CCCCN4CCN(CC4)C5=NC=CC=N5.Cl
C21H30ClN5O2
ACVFJYKNBOHIMH-DPFKZJTMSA-N
13422623

Specifications

Tandospirone hydrochloride
99095-10-0
tandospirone hydrochloride, unii-rwr6slb9p6, rwr6slb9p6, metanopirone, sediel, tandospirone hydrochloride mi, 4,7-methano-1h-isoindole-1,3 2h-dione, hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-, hydrochloride 1:1 , 3ar,4s,7r,7as-rel, 3ar,4s,7r,7as-rel-hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-4,7-methano-1h-isoindole-1,3 2h-dione hydrochloride, 4,7-methano-1h-isoindole-1,3 2h-dione, hexahydro-2-4-4-2-pyrimidinyl-1-piperazinyl butyl-, hydrochloride, 3ar,4s,7r,7as-rel
C1CC2CC1C3C2C(=O)N(C3=O)CCCCN4CCN(CC4)C5=NC=CC=N5.Cl
13422623
>98%
50mg
C21H30ClN5O2
ACVFJYKNBOHIMH-DPFKZJTMSA-N
419.954
419.95
Videos