missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Tambour door cupboard, Ekwo, grey,/black, 1050mm x 1000mm x 450mm
Brand: Manutan A106941
Code : Z3
Additional Details : Weight : 37.90000kg
Description
Sliding doors allow 100% access without doors being swung open. Shelves adjustable in 20mm increments. Lockable doors, supplied with two keys from a mastered series.- Number of shelves: 2
- Dimensions (H x W x D): 1050mm x 1000mm x 450mm
- Grey/black