missing translation for 'onlineSavingsMsg'
Learn More

Talopram hydrochloride, Tocris Bioscience™

Potent, selective inhibitor of noradrenalin transporters

Brand:  RD Systems (Bio-Techne) 3145/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13462524

  • £145.80 / 10mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

Potent, selective inhibitor of the noradrenalin transporter (NET) (IC50 = 2.9 nM). Exhibits selectivity for NET against SERT (5-HT transporters) and DAT (dopamine transporters). Displays a similar structure but different pharmacological profile to citalopram (Cat. No. 1427).
Specifications

Chemical Identifiers

7013-41-4
331.884
talopram hydrochloride, talopram hcl, phtalapromine hydrochloride, talopram hydrochloride usan, n,3,3-trimethyl-1-phenyl-1-phthalanpropylamine hydrochloride, 1-phthalanpropylamine, 1-phenyl-n,3,3-trimethyl-, hydrochloride, 1-phthalanpropylamine, n,3,3-trimethyl-1-phenyl-, hydrochloride, phthalan, 3,3-dimethyl-1-3-methylaminopropyl-1-phenyl-, hydrochloride, 1-isobenzofuranpropanamine, 1,3-dihydro-n,3,3-trimethyl-1-phenyl-, hydrochloride, talopram hydrochloride usan
3-(3,3-dimethyl-1-phenyl-2-benzofuran-1-yl)-N-methylpropan-1-amine;hydrochloride
C20H26ClNO
JZXJIRQPHHWYGC-UHFFFAOYSA-N
71176
CC1(C2=CC=CC=C2C(O1)(CCCNC)C3=CC=CC=C3)C.Cl

Specifications

Talopram hydrochloride
7013-41-4
talopram hydrochloride, talopram hcl, phtalapromine hydrochloride, talopram hydrochloride usan, n,3,3-trimethyl-1-phenyl-1-phthalanpropylamine hydrochloride, 1-phthalanpropylamine, 1-phenyl-n,3,3-trimethyl-, hydrochloride, 1-phthalanpropylamine, n,3,3-trimethyl-1-phenyl-, hydrochloride, phthalan, 3,3-dimethyl-1-3-methylaminopropyl-1-phenyl-, hydrochloride, 1-isobenzofuranpropanamine, 1,3-dihydro-n,3,3-trimethyl-1-phenyl-, hydrochloride, talopram hydrochloride usan
CC1(C2=CC=CC=C2C(O1)(CCCNC)C3=CC=CC=C3)C.Cl
331.884
331.88
10mg
C20H26ClNO
JZXJIRQPHHWYGC-UHFFFAOYSA-N
3-(3,3-dimethyl-1-phenyl-2-benzofuran-1-yl)-N-methylpropan-1-amine;hydrochloride
71176
>99%
Videos