missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550838
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total Talin protein.
Talin 1 Polyclonal antibody specifically detects Talin 1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Talin 1 | |
Polyclonal | |
Unconjugated | |
TLN1 | |
ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
21894, 7094 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.8 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P26039, Q9Y490 | |
TLN1 | |
Synthetic peptide corresponding to residues near the N terminal of human talin 1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |