missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, Powder, Multi-Compendial, J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Quantity | 500 g |
Grade | Multi-Compendial |
Physical Form | Powder |
Chemical Name or Material | Talc |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'