missing translation for 'onlineSavingsMsg'
Learn More

TAK 715, Tocris Bioscience™

Potent p38 MAPK inhibitor; anti-inflammatory

Brand:  Tocris (Bio-Techne) 425450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444444

  • £633.57 / 50mg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Inhibitor of p38 MAPK (IC50 = 7.1 nM for p38 MAPKα). Also inhibits Wnt-3a-stimulated β-catenin signaling. Inhibits 22 kinases by more than 80%, including CK1δ/ε. Anti-inflammatory and anti-rheumatoid arthritis agent. Orally bioavailable.
Specifications

Chemical Identifiers

303162-79-0
399.512
unii-we92u03c5z, n-4-2-ethyl-4-m-tolyl thiazol-5-yl pyridin-2-yl benzamide, n-4-2-ethyl-4-3-methylphenyl-1,3-thiazol-5-yl pyridin-2-yl benzamide, n-4-2-ethyl-4-m-tolylthiazol-5-yl pyridin-2-yl benzamide, benzamide, n-4-2-ethyl-4-3-methylphenyl-5-thiazolyl-2-pyridinyl, kinome_2655, d0o6zo, n-4-2-ethyl-4-3-methylphenyl-1,3-thiazol-5-yl-2-pyridyl benzamide
N-[4-[2-ethyl-4-(3-methylphenyl)-1,3-thiazol-5-yl]pyridin-2-yl]benzamide
C24H21N3OS
HEKAIDKUDLCBRU-UHFFFAOYSA-N
9952773
CCC1=NC(=C(S1)C2=CC(=NC=C2)NC(=O)C3=CC=CC=C3)C4=CC(=CC=C4)C

Specifications

50mg
303162-79-0
unii-we92u03c5z, n-4-2-ethyl-4-m-tolyl thiazol-5-yl pyridin-2-yl benzamide, n-4-2-ethyl-4-3-methylphenyl-1,3-thiazol-5-yl pyridin-2-yl benzamide, n-4-2-ethyl-4-m-tolylthiazol-5-yl pyridin-2-yl benzamide, benzamide, n-4-2-ethyl-4-3-methylphenyl-5-thiazolyl-2-pyridinyl, kinome_2655, d0o6zo, n-4-2-ethyl-4-3-methylphenyl-1,3-thiazol-5-yl-2-pyridyl benzamide
CCC1=NC(=C(S1)C2=CC(=NC=C2)NC(=O)C3=CC=CC=C3)C4=CC(=CC=C4)C
399.512
399.51
TAK 715
C24H21N3OS
HEKAIDKUDLCBRU-UHFFFAOYSA-N
N-[4-[2-ethyl-4-(3-methylphenyl)-1,3-thiazol-5-yl]pyridin-2-yl]benzamide
9952773
>99%
Videos
Documents