missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560968
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KGPNQGRLVP RLSVGAVSSK PTT Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 96%.
TAF9B Polyclonal antibody specifically detects TAF9B in Human samples. It is validated for Immunohistochemistry.Specifications
TAF9B | |
Polyclonal | |
Unconjugated | |
Taf9b | |
BC066223; Dn7; DN-7; neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; neuronal cell death-related protein 7; Taf9b; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAF9L; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAFII31; TAFII31L; TATA box binding protein associated factor 9b; TATA-box binding protein associated factor 9b; TBP-associated factor 9L; TFIID-31; transcription associated factor TAFII31L; transcription initiation factor IID, 31kD subunit; Transcription initiation factor TFIID subunit 9B; Transcription initiation factor TFIID subunit 9-li | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HBM6 | |
Taf9b | |
Recombinant protein corresponding to Human TAF9B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only