missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CD267 (TACI) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0097
Code : NEW
Additional Details : Weight : 0.01000kg
Description
TACI Synthetic Peptide, PEP-0097, from Invitrogen.
This peptide corresponds to 15 amino amino terminus of human TACI.PEP-0097 can be used as a blocking peptide with polyclonal antibody PA5-19972.Specifications
23495 | |
purified | |
Human CD267 (TACI) | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O14836 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
TNFRSF13B | |
0.02% sodium azide | |
50 μg | |
PBS |