missing translation for 'onlineSavingsMsg'
Learn More

t-Butyl4-[4-(ethoxycarbonyl)-1,3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate, ≥95%, Maybridge

Brand:  Maybridge CC31023DA

Code : 31

Additional Details : CAS Number : 365413-31-6 Weight : 0.05100kg

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10527853

  • £92.00 / 1g


Estimated Shipment date
from Supplier 23-07-2018
Add to basket

Specifications

Chemical Identifiers

Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
96.5
97%
C16H24N2O4S
MFCD06659069
365413-31-6
100.0
97%
340.44

Specifications

Tert-butyl 4-[4-(ethoxycarbonyl)-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
96.5
Amber Glass Bottle
1g
C16H24N2O4S
MFCD06659069
365413-31-6
100.0
97%
97%
340.44
Videos
SDS