missing translation for 'onlineSavingsMsg'
Learn More

T 0901317, Tocris Bioscience™

Potent liver X receptor (LXR) agonist; also CAR inverse agonist

Brand:  Tocris (Bio-Techne) 237310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13451104

  • £108.78 / 10mg

Estimated Shipment date
from Supplier 21-02-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent, high affinity liver X receptor (LXR) agonist (EC50 ∽ 50 nM, Kd values are 7 and 22 nM for LXR-α and LXR-β respectively). Upregulates expression of the ABCA1 gene associated with cholesterol efflux regulation and HDL metabolism. Decreases amyloid-β production in primary neurons in vitro. Displays an EC50 of ∽ 5 μM for activation of bile acid farnesoid X receptors (FXRs); 10-fold more potent than natural FXR ligand chenodeoxycholic acid. Also exhibits inverse agonist activity at constitutive androstane receptors (CAR).
Specifications

Chemical Identifiers

293754-55-9
481.333
n-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, chembl62136, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethylethyl phenyl sulfonamide, 1pqc, d0b5fc, n-4-1,1,1,3,3,3-hexafluoro-2-hydroxy-propan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, t, benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl
CHEBI:39976
C1=CC=C(C=C1)S(=O)(=O)N(CC(F)(F)F)C2=CC=C(C=C2)C(C(F)(F)F)(C(F)(F)F)O
C17H12F9NO3S
SGIWFELWJPNFDH-UHFFFAOYSA-N
447912
N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide

Specifications

10mg
293754-55-9
n-4-1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, chembl62136, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethylethyl phenyl sulfonamide, 1pqc, d0b5fc, n-4-1,1,1,3,3,3-hexafluoro-2-hydroxy-propan-2-yl phenyl-n-2,2,2-trifluoroethyl benzenesulfonamide, t, benzenesulfonamide, n-2,2,2-trifluoroethyl-n-4-2,2,2-trifluoro-1-hydroxy-1-trifluoromethyl ethyl phenyl
C1=CC=C(C=C1)S(=O)(=O)N(CC(F)(F)F)C2=CC=C(C=C2)C(C(F)(F)F)(C(F)(F)F)O
481.333
CHEBI:39976
>98%
T 0901317
C17H12F9NO3S
SGIWFELWJPNFDH-UHFFFAOYSA-N
N-[4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl]-N-(2,2,2-trifluoroethyl)benzenesulfonamide
447912
481.33
Videos
Documents