Learn More
T 0070907, Tocris Bioscience™
Highly potent and selective PPARγ antagonist
Brand:  Tocris (Bio-Techne) 230150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13440984
  • £565.95 / 50mg
Estimated Shipment date
from Supplier 27-03-2020
Description
Description
Potent and selective PPARγ antagonist (IC50 = 1 nM). Displays > 800-fold selectivity for PPARγ over PPARα and PPARδ. Blocks transcriptional activity of PPARγ in vitro and inhibits rosiglitazone-induced adipogenesis.
Specifications
Chemical Identifiers
313516-66-4
277.664
2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
2-chloro-5-nitro-N-pyridin-4-ylbenzamide
C12H8ClN3O3
FRPJSHKMZHWJBE-UHFFFAOYSA-N
2777391
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl
Specifications
50mg
313516-66-4
2-chloro-5-nitro-n-4-pyridinylbenzamide, 2-chloro-5-nitro-n-pyridin-4-yl benzamide, 2-chloro-5-nitro-n-4-pyridyl benzamide, benzamide, 2-chloro-5-nitro-n-4-pyridinyl, 2-chloro-5-nitro-n-4-pyridinyl-benzamide, d03bpg, d08gqc, 2-chloro-5-nitro-n-4-pyridinyl benzamide, benzamide,2-chloro-5-nitro-n-4-pyridinyl, n1-4-pyridyl-2-chloro-5-nitrobenzamide
C1=CC(=C(C=C1[N+](=O)[O-])C(=O)NC2=CC=NC=C2)Cl
277.664
277.67
T 0070907
C12H8ClN3O3
FRPJSHKMZHWJBE-UHFFFAOYSA-N
2-chloro-5-nitro-N-pyridin-4-ylbenzamide
2777391
>99%