missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hamilton™ Syringe
Brand: Hamilton™ 200743
Code : 13
Additional Details : Weight : 0.02500kg
Specifications
0.23 mm | |
Syringe | |
45° |
75 mm | |
5 μL |
Brand: Hamilton™ 200743
Code : 13
Additional Details : Weight : 0.02500kg
0.23 mm | |
Syringe | |
45° |
75 mm | |
5 μL |