missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ CTC PAL HPLC Fixed Needle Autosampler Syringe
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
Brand: Trajan™ 003715
Code : 16
Additional Details : Weight : 0.10000kg
Description
- High temperature syringes for headspace application with temperatures up to 150°C
- PTFE plunger seal option for smooth, leak free operation - suitable for both liquid and gas samples, with replaceable plungers
- Specifications of the syringes are exactly matched to CTC holders to ensure proper fit and reliable functioning
Specifications
PTFE (Plunger) | |
0.17 mm | |
Gastight | |
25 μL |
22 G | |
0.72 mm | |
25F-CTC-GT-LC | |
51 mm |